Pentafluorophenyl-4-nitrobenzenesulphonate ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Pentafluorophenyl 4-Nitrobenzenesulphonate
P2231-1G
P2231-5G
TCIAP2231-1GEA
405
NOK
TCIAP2231-1G
TCIAP2231-5G
Pentafluorophenyl-4-nitrobenzenesulphonate ≥98.0% (by GC)
Pentafluorophenyl-4-nitrobenzenesulphonate
Formel:
C₁₂H₄F₅NO₅S MW: 369,2209 g/mol |
MDL nummer:
MFCD05975120 CAS-nummer: 244633-31-6 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...