MOPS
Leverandør: RESEARCH ORGANICS
RES9615M-A102X
RES9615M-A103X
RESERES9615M-A102XEA
0
NOK
RESERES9615M-A102X
RESERES9615M-A103X
MOPS
MOPS
Formel:
C₇H₁₅NO₄S Smeltepunkt: > 250 °C Flash punkt: 110 °C (closed cup) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00006183 CAS-nummer: 1132-61-2 EINECS: 214-478-5 Merck Indeks: 13,06288 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...