1-Phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-benzimidazole ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
1-Phenyl-2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-benzo[d]imidazole, 3-(1-Phenyl-1H-benzimidazole-2-yl)phenylboronic acid Pinacol ester
P2263-1G
P2263-5G
TCIAP2263-1GEA
1000
NOK
TCIAP2263-1G
TCIAP2263-5G
1-Phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-benzimidazole ≥98.0% (by GC, titration analysis)
1-Phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-benzimidazole
Formel:
C₂₅H₂₅BN₂O₂ MW: 396,30 g/mol |
MDL nummer:
MFCD16038225 CAS-nummer: 952514-86-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 172.0 to 176.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...