Piperidine-2,4-dione ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2,4-Piperidinedione, 2,4-Dioxopiperidine

P2296-200MG P2296-1G
TCIAP2296-200MGEA 260 NOK
TCIAP2296-200MG TCIAP2296-1G
Piperidine-2,4-dione ≥98.0% (by GC, titration analysis)
Piperidine-2,4-dione
Formel: C₅H₇NO₂
MW: 113,1146 g/mol
MDL nummer: MFCD08704814
CAS-nummer: 50607-30-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR