4-(2-Propynyloxy)benzenecarbaldehyde ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
4-(Prop-2-yn-1-yloxy)benzaldehyde, 4-(2-Propynyloxy)benzaldehyde, 4-(Propargyloxy)benzaldehyde
P2339-200MG
P2339-1G
TCIAP2339-200MGEA
785
NOK
TCIAP2339-200MG
TCIAP2339-1G
4-(2-Propynyloxy)benzenecarbaldehyde ≥98.0% (by GC)
4-(2-Propynyloxy)benzenecarbaldehyde
Formel:
C₁₀H₈O₂ MW: 160,17 g/mol |
MDL nummer:
MFCD01088150 CAS-nummer: 5651-86-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...