4'-Pentyloxybiphenyl-4-boronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
Synonymer:
[4'-(Pentyloxy)-[1,1'-biphenyl]-4-yl]boronic acid, (4'-(Pentyloxy)-[1,1'-biphenyl]-4-yl)boronic acid, 4'-Amyloxybiphenyl-4-boronic acid
P2365-1G
P2365-5G
TCIAP2365-1GEA
675
NOK
TCIAP2365-1G
TCIAP2365-5G
4'-Pentyloxybiphenyl-4-boronic acid (contains varying amounts of Anhydride)
4'-Pentyloxybiphenyl-4-boronic acid
Formel:
C₁₇H₂₁BO₃ MW: 284,16 g/mol Smeltepunkt: 197°C |
MDL nummer:
MFCD07644475 CAS-nummer: 158937-25-8 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 107.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...