2-Phenyl-9H-carbazole ≥98.0% (by GC)

Leverandør: TCI
Warning

P2375-1G P2375-5G
TCIAP2375-1GEA 525 NOK
TCIAP2375-1G TCIAP2375-5G
2-Phenyl-9H-carbazole ≥98.0% (by GC)
2-Phenyl-9H-carbazole
Formel: C₁₈H₁₃N
MW: 243,31 g/mol
MDL nummer: MFCD26402145
CAS-nummer: 88590-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4235663ae3fc2eac4d0c75920e922f7ea2de7c01&vid=4235663ae3fc2eac4d0c75920e922f7ea2de7c01&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR