2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4,4,5,5-Tetramethyl-2-(6-phenyl-3-pyridyl)-1,3,2-dioxaborolane, 6-Phenylpyridine-3-boronic acid pinacol ester, 6-Phenyl-3-pyridylboronic acid Pinacol ester
P2384-200MG
P2384-1G
TCIAP2384-200MGEA
1075
NOK
TCIAP2384-200MG
TCIAP2384-1G
2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥98.0% (by GC, titration analysis)
2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formel:
C₁₇H₂₀BNO₂ MW: 281,16 g/mol |
MDL nummer:
MFCD12032565 CAS-nummer: 879291-27-7 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...