PhenoFluor™ Mix

Leverandør: TCI
Danger

Synonymer: PhenoFluor™ Mix, (2-Chloro-1,3-bis(2,6-diisopropylphenyl)imidazolium chloride)

P2420-1G P2420-5G
TCIAP2420-1GEA 1875 NOK
TCIAP2420-1G TCIAP2420-5G
PhenoFluor™ Mix
2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazol-3-ium chloride cesium fluoride complex
Formel: C₂₇H₃₆Cl₂CsFN₂
MW: 611,39 g/mol
MDL nummer: MFCD29472387
CAS-nummer: 1648825-53-9
FN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2322e3f6574cacd6d1599f0b068097b0f04a32d1&vid=2322e3f6574cacd6d1599f0b068097b0f04a32d1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Assay 30.0 to 32.0w% [as 2-Chloro-1,3-bis (2,6-diisopropylphenyl)-1H-imidazolium Chloride]

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR