1-(4-Cyanophenyl)piperazine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-(Piperazin-1-yl)benzonitrile, 4-(1-Piperazinyl)benzonitrile, 1-(4-Cyanophenyl)piperazine

P2449-1G P2449-5G
TCIAP2449-1GEA 1075 NOK
TCIAP2449-1G TCIAP2449-5G
1-(4-Cyanophenyl)piperazine ≥98.0% (by GC, titration analysis)
1-(4-Cyanophenyl)piperazine
Formel: C₁₁H₁₃N₃
MW: 187,2410 g/mol
MDL nummer: MFCD01320890
CAS-nummer: 68104-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=524c4c8f7c941d10913dbd98e9ee916a4db77a7d&vid=524c4c8f7c941d10913dbd98e9ee916a4db77a7d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR