4-(1-Pyrazolyl)benzonitrile ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
1-(4-Cyanophenyl)pyrazole
P2450-1G
P2450-5G
TCIAP2450-1GEA
460
NOK
TCIAP2450-1G
TCIAP2450-5G
4-(1-Pyrazolyl)benzonitrile ≥98.0% (by GC)
4-(1H-Pyrazol-3-yl)benzonitrile
Formel:
C₁₀H₇N₃ MW: 169,19 g/mol |
MDL nummer:
MFCD09033533 CAS-nummer: 25699-83-6 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...