Pirenzepine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
11-(2-(4-Methylpiperazin-1-yl)acetyl)-5,11-dihydro-6H-benzo[e]pyrido[3,2-b][1,4]diazepin-6-one dihydrochloride
P2457-1G
P2457-200MG
TCIAP2457-1GEA
1800
NOK
TCIAP2457-1G
TCIAP2457-200MG
Pirenzepine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Pirenzepine dihydrochloride
Formel:
C₁₉H₂₁N₅O₂·2HCl MW: 424,32 g/mol |
MDL nummer:
MFCD00055214 CAS-nummer: 29868-97-1 EINECS: 249-907-5 Merck Indeks: 14,07491 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | max. 4.9 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...