1-Phenyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
1-Phenylpyrazole-4-carboxaldehyde, 1-Phenyl-1H-pyrazole-4-carboxaldehyde
P2473-1G
P2473-5G
TCIAP2473-1GEA
960
NOK
TCIAP2473-1G
TCIAP2473-5G
1-Phenyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
1-Phenyl-1H-pyrazole-4-carbaldehyde
Formel:
C₁₀H₈N₂O MW: 172,18 g/mol |
MDL nummer:
MFCD02179568 CAS-nummer: 54605-72-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...