Propylamine hydrobromide ≥97.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Warning

Synonymer: Propylammonium bromide

P2502-1G P2502-5G
TCIAP2502-1GEA 245 NOK
TCIAP2502-1G TCIAP2502-5G
Propylamine hydrobromide ≥97.0% (by total nitrogen and titration analysis)
Propylamine hydrobromide
Formel: C₃H₉N·HBr
Smeltepunkt: 182°C
MDL nummer: MFCD30749306
CAS-nummer: 4905-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae4bff3ee6c3c96955076bb0e6e0737d37261c38&vid=ae4bff3ee6c3c96955076bb0e6e0737d37261c38&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR