Potassium tris(trifluoromethanesulphonyl)methanide ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Danger
|
P2520-1G
P2520-5G
TCIAP2520-1GEA
2000
NOK
TCIAP2520-1G
TCIAP2520-5G
Potassium tris(trifluoromethanesulphonyl)methanide ≥98.0% (by titrimetric analysis)
Potassium tris(trifluoromethanesulphonyl)methanide
Formel:
C₄F₉KO₆S₃ MW: 450,30 g/mol |
MDL nummer:
MFCD18072174 CAS-nummer: 114395-69-6 FN: 3261 ADR: 8,II |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...