Pilsicainide hydrochloride ≥98.0% (by HPLC)

Leverandør: TCI
Danger

Synonymer: N-(2,6-Dimethylphenyl)tetrahydro-1H-pyrrolizine-7a(5H)-acetamide hydrochloride

P2634-250MG P2634-50MG
TCIAP2634-250MGEA 2600 NOK
TCIAP2634-250MG TCIAP2634-50MG
Pilsicainide hydrochloride ≥98.0% (by HPLC)
Pilsicainide hydrochloride
Formel: C₁₇H₂₄N₂O·HCl
MW: 308,85 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00903769
CAS-nummer: 88069-49-2
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,07427

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 97.0 %
Melting point 211.0 to 215.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR