Pentachlorobenzonitrile ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,3,4,5,6-Pentachlorobenzonitrile

P2643-25G P2643-5G
TCIAP2643-25GEA 2725 NOK
TCIAP2643-25G TCIAP2643-5G
Pentachlorobenzonitrile ≥98.0% (by GC)
Pentachlorobenzonitrile
Formel: C₇Cl₅N
MDL nummer: MFCD00173666
CAS-nummer: 20925-85-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 215.0 to 219.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR