Potassium(4-chlorophenyl)trifluoroborate ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: (4-Chlorophenyl)trifluoroboric acid potassium salt, Potassium (4-chlorophenyl)trifluoroborate

P2654-25G P2654-5G
TCIAP2654-25GEA 3100 NOK
TCIAP2654-25G TCIAP2654-5G
Potassium(4-chlorophenyl)trifluoroborate ≥98.0% (by HPLC)
Potassium(4-chlorophenyl)trifluoroborate
Formel: C₆H₄BClF₃K
MW: 218,45 g/mol
MDL nummer: MFCD04115770
CAS-nummer: 661465-44-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR