2-Phenylethyl n-Octanoate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: n-Octanoic acid 2-Phenylethyl ester, Phenethyl n-Octanoate, n-Octanoic acid Phenethyl ester

P2741-100G P2741-25G
TCIAP2741-100GEA 1175 NOK
TCIAP2741-100G TCIAP2741-25G
2-Phenylethyl n-Octanoate ≥98.0% (by GC)
2-Phenylethyl n-Octanoate
Formel: C₁₆H₂₄O₂
MW: 248,37 g/mol
Kokepunkt: 296°C
MDL nummer: MFCD00209536
CAS-nummer: 5457-70-5
EINECS: 226-718-6

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR