Polypropylene Glycol Diacrylate n=∼12 stabilised
Leverandør: TCI
|
Warning
|
Synonymer:
PPG Diacrylate, PPGDA
Stabilisering:
Stabilised with 4-methoxyphenol (MEHQ)
P2759-25G
P2759-500G
TCIAP2759-25GEA
220
NOK
TCIAP2759-25G
TCIAP2759-500G
Polypropylene Glycol Diacrylate n=∼12 stabilised
Polypropylene Glycol Diacrylate
Lagringstemperatur:
Kjøleskap |
MDL nummer:
MFCD00081912 CAS-nummer: 52496-08-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
The average value of n | 11.0 to 13.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...