Polyethylene glycol dimethacrylate n=∼14 stabilised

Leverandør: TCI
Warning

Synonymer: PEGDMA, Polyethylene Glycol Dimethacrylate average Mn 550, PEG dimethacrylate, Polyethylene Glycol Dimethacrylate

Stabilisering: Stabilised with 4-methoxyphenol (MEHQ)

P2760-25G P2760-500G
TCIAP2760-25GEA 260 NOK
TCIAP2760-25G TCIAP2760-500G
Polyethylene glycol dimethacrylate n=∼14 stabilised
Polyethylene glycol dimethacrylate
Formel: (C₂H₄O)nC₈H₁₀O₃
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00014932
CAS-nummer: 25852-47-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
The average value of n 13.0 to 15.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR