Rebaudioside A ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: (2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl (4R,4aS,6aR,9S,11aR,11bS)-9-(((2S,3R,4S,5R,6R)-5-hydroxy-6-(hydroxymethyl)-3,4-bis(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-4,11b-dimethyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylate

R0095-25G R0095-5G
TCIAR0095-25GEA 1350 NOK
TCIAR0095-25G TCIAR0095-5G
Rebaudioside A ≥98.0% (by HPLC)
Rebaudioside A
Formel: C₄₄H₇₀O₂₃
MW: 967,02 g/mol
Smeltepunkt: 235°C
MDL nummer: MFCD02183463
CAS-nummer: 58543-16-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Specific rotation [a]20/D -30.0 to -36.0 deg (C=1、H2O) (calcd.on dried substance)
Drying loss max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR