Rabeprazole sodium salt ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: Sodium 2-(((4-(3-methoxypropoxy)-3-methylpyridin-2-yl)methyl)sulphinyl)benzo[d]imidazol-1-ide

R0115-100MG R0115-1G
TCIAR0115-100MGEA 245 NOK
TCIAR0115-100MG TCIAR0115-1G
Rabeprazole sodium salt ≥98.0% (by HPLC)
Rabeprazole sodium salt
Formel: C₁₈H₂₁N₃O₃S
MW: 381,43 g/mol
Smeltepunkt: 150°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD02092688
CAS-nummer: 117976-90-6
Merck Indeks: 14,08089
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=192c1e1b33e6f4eb6955152cd2940ad5d4e3219d&vid=192c1e1b33e6f4eb6955152cd2940ad5d4e3219d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR