RuCl2[(S)-xylbinap][(S,S)-dpen]
Leverandør: TCI
Synonymer:
Dichloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) RuCl2[(S)-xylbinap][(S,S)-dpen], Dichloro[(S)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl][(S,S)-1,2-diphenylethylenediamine]ruthenium(II)
R0134-200MG
R0134-1G
TCIAR0134-200MGEA
525
NOK
TCIAR0134-200MG
TCIAR0134-1G
RuCl2[(S)-xylbinap][(S,S)-dpen]
RuCl2[(S)-xylbinap][(S,S)-dpen]
Formel:
C₆₆H₆₄Cl₂N₂P₂Ru MW: 1119,17 g/mol |
MDL nummer:
MFCD09753030 CAS-nummer: 220114-03-4 |
Spesifikasjon Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...