Roxithromycin ≥95.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: (3R,4S,5S,6R,7R,9R,11S,12R,13S,14R,E)-6-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-10-(((2-methoxyethoxy)methoxy)imino)-3,5,7,9,11,13-hexamethyloxacyclotetradecan-2-one

R0164-5G R0164-25G
TCIAR0164-5GEA 595 NOK
TCIAR0164-5G TCIAR0164-25G
Roxithromycin ≥95.0% (by HPLC)
Roxithromycin
Formel: C₄₁H₇₆N₂O₁₅
MW: 837,04 g/mol
Smeltepunkt: 120°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00214389
CAS-nummer: 80214-83-1
Merck Indeks: 14,08276
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f259f1446a7ee8946095fec0b97cdab5c25abead&vid=f259f1446a7ee8946095fec0b97cdab5c25abead&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 3.10 to 3.60 %
Specific rotation [a]20/D -93.0 to -96.0 deg (C=1, Acetone) (calcd.on anh.substance)
Water max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR