Repaglinide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]benzoic acid
R0179-200MG
R0179-1G
TCIAR0179-200MGEA
795
NOK
TCIAR0179-200MG
TCIAR0179-1G
Repaglinide ≥98.0% (by HPLC, titration analysis)
Repaglinide
Formel:
C₂₇H₃₆N₂O₄ MW: 452,59 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00906179 CAS-nummer: 135062-02-1 Merck Indeks: 14,08136 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Specific rotation [a]20/D | +7.0 to +10.0 deg (C=1,MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...