Monosodium 5-sulphoisophthalate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Sodium 3,5-dicarboxybenzenesulphonate, 5-Sulphoisophthalic acid monosodium salt, Sodium 5-sulphoisophthalate

S0148-25G S0148-500G S0148-100G
TCIAS0148-25GEA 220 NOK
TCIAS0148-25G TCIAS0148-500G TCIAS0148-100G
Monosodium 5-sulphoisophthalate ≥98.0% (by titrimetric analysis)
Monosodium 5-sulphoisophthalate
Formel: C₈H₆O₇S
MW: 268,17 g/mol
MDL nummer: MFCD00007495
CAS-nummer: 6362-79-4
EINECS: 228-845-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7eff7a7851043b7ba58d14d640b2dc8fb826cf9&vid=f7eff7a7851043b7ba58d14d640b2dc8fb826cf9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR