2,2'-sulphonylbis(4-tert-octylphenol) ≥96.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Bis[2-hydroxy-5-(1,1,3,3-tetramethylbutyl)phenyl] sulphone, Bis(2-hydroxy-5-tert-octylphenyl) Sulphone
S0478-5G
S0478-1G
TCIAS0478-5GEA
2725
NOK
TCIAS0478-5G
TCIAS0478-1G
2,2'-sulphonylbis(4-tert-octylphenol) ≥96.0% (by GC, titration analysis)
2,2'-sulphonylbis(4-tert-octylphenol)
Formel:
C₂₈H₄₂O₄S MW: 474,70 g/mol |
MDL nummer:
MFCD00026328 CAS-nummer: 15452-89-8 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 144.0 to 148.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...