6'-Sialyllactose sodium salt ≥98.0% (by HPLC)

Leverandør: TCI

S0886-20MG S0886-100MG
TCIAS0886-20MGEA 1850 NOK
TCIAS0886-20MG TCIAS0886-100MG
6'-Sialyllactose sodium salt ≥98.0% (by HPLC)
6'-Sialyllactose sodium salt
Formel: C₂₃H₃₈NNaO₁₉
Lagringstemperatur: Fryseskap
CAS-nummer: 157574-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02de28b66d1e3ff62a48cbd24e8caa7ffc784ddb&vid=02de28b66d1e3ff62a48cbd24e8caa7ffc784ddb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR