Stiripentol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 4,4-Dimethyl-1-[3,4(methylenedioxy)-phenyl]-1-penten-3-ol, 1-(1,3-Benzodioxol-5-yl)-4,4-dimethyl-1-penten-3-ol, 4,4-Dimethyl-1-(3,4-methylenedioxyphenyl)-1-penten-3-ol, 1-(Benzo[d][1,3]dioxol-5-yl)-4,4-dimethylpent-1-en-3-ol

S0905-1G S0905-5G
TCIAS0905-1GEA 2125 NOK
TCIAS0905-1G TCIAS0905-5G
Stiripentol ≥98.0% (by GC)
Stiripentol
Formel: C₁₄H₁₈O₃
MW: 234,30 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00869310
CAS-nummer: 49763-96-4
EINECS: 256-480-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bc858ccbbb4a2dc952d718b385659e7d004e283&vid=7bc858ccbbb4a2dc952d718b385659e7d004e283&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR