Biotin-PEG2-NHS
Leverandør: TCI
Synonymer:
N-Succinimidyl 9-(Biotinamido)-4,7-dioxanonanoate, 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(5-(2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)ethoxy)ethoxy)propanoate, Biotin-PEG2-NHS ester, 9-(Biotinamido)-4,7-dioxanonanoic acid N-Succinimidyl ester
S0955-25MG
S0955-100MG
TCIAS0955-25MGEA
1300
NOK
TCIAS0955-25MG
TCIAS0955-100MG
Biotin-PEG2-NHS
Biotin-PEG2-NHS
Formel:
C₂₁H₃₂N₄O₈S MW: 500,57 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD00467154 CAS-nummer: 596820-83-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Elemental analysis (Carbon) | 47.90 to 52.90 % |
Elemental analysis (Nitrogen) | 10.60 to 11.80 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...