AMAS (N-Succinimidyl maleimidoacetate)

Leverandør: TCI

Synonymer: AMAS

S0968-100MG S0968-20MG
TCIAS0968-100MGEA 2925 NOK
TCIAS0968-100MG TCIAS0968-20MG
AMAS (N-Succinimidyl maleimidoacetate)
AMAS (N-Succinimidyl maleimidoacetate)
Formel: C₁₀H₈N₂O₆
MW: 252,18 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00133523
CAS-nummer: 55750-61-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Elemental analysis (Nitrogen) 10.50 to 11.50 %
Melting point 186.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR