Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulphonate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: Sodium 5-Isopropyl-3,8-dimethylazulene-1-sulphonate, Sodium Guaiazulenesulphonate, 7-Isopropyl-1,4-dimethyl-azulene-3-sulphonic acid sodium salt, 5-Isopropyl-3,8-dimethylazulene-1-sulphonic acid sodium salt, Guaiazulenesulphonic acid sodium salt, Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulphonate

S0976-1G S0976-5G
TCIAS0976-1GEA 850 NOK
TCIAS0976-1G TCIAS0976-5G
Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulphonate ≥98.0% (by HPLC, titration analysis)
Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulphonate
Formel: C₁₅H₁₇NaO₃S
MW: 300,35 g/mol
Smeltepunkt: 98°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00866167
CAS-nummer: 6223-35-4
EINECS: 228-309-8
Merck Indeks: 14,04554
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cf237c0619648a719a4672241d13b54cf5cde6f&vid=8cf237c0619648a719a4672241d13b54cf5cde6f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Blue to Dark blue powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 2.5 to 6.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR