Sodium 2-Thiobarbiturate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 2-Thiobarbituric acid sodium salt

T0193-500G T0193-25G
TCIAT0193-500GEA 4200 NOK
TCIAT0193-500G TCIAT0193-25G
Sodium 2-Thiobarbiturate ≥98.0% (by titrimetric analysis)
Sodium 2-Thiobarbiturate
Formel: C₄H₃N₂NaO₂S
MW: 166,13 g/mol
MDL nummer: MFCD00065054
CAS-nummer: 31645-12-2
EINECS: 250-749-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR