L-Tyrosine ethyl ester ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Ethyl L-tyrosinate, H-Tyr-OEt

T0551-1G T0551-5G
TCIAT0551-1GEA 310 NOK
TCIAT0551-1G TCIAT0551-5G
L-Tyrosine ethyl ester ≥98.0% (by titrimetric analysis)
L-Tyrosine ethyl ester
Formel: C₁₁H₁₅NO₃
MW: 209,25 g/mol
MDL nummer: MFCD00063046
CAS-nummer: 949-67-7
EINECS: 213-442-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 102.0 to 105.0 deg-C
Specific rotation [a]20/D +16.0 to +19.0 deg (C=5, EtOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR