Diisodecyl-4-cyclohexene-1,2-dicarboxylate ≥97.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Diisodecyl 1,2,3,6-Tetrahydrophthalate, 4-Cyclohexene-1,2-dicarboxylic acid diisodecyl ester, 1,2,3,6-Tetrahydrophthalic acid diisodecyl ester

T0603-25ML T0603-500ML
TCIAT0603-25MLEA 350 NOK
TCIAT0603-25ML TCIAT0603-500ML
Diisodecyl-4-cyclohexene-1,2-dicarboxylate ≥97.0% (by titrimetric analysis)
Diisodecyl-4-cyclohexene-1,2-dicarboxylate
Formel: C₂₈H₅₀O₄
MW: 450,70 g/mol
MDL nummer: MFCD00070615
CAS-nummer: 87826-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3fb5429f13204eea834f2332a4e26662344ffc0&vid=a3fb5429f13204eea834f2332a4e26662344ffc0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Ester Value) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR