Thymoquinone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: p-Mentha-3,6-diene-2,5-dione, 2-Isopropyl-5-methylcyclohexa-2,5-diene-1,4-dione, 5-Isopropyl-2-methyl-1,4-benzoquinone

T0795-1G T0795-5G
TCIAT0795-1GEA 525 NOK
TCIAT0795-1G TCIAT0795-5G
Thymoquinone ≥98.0% (by GC)
Thymoquinone
Formel: C₁₀H₁₂O₂
MW: 164,20 g/mol
Kokepunkt: 232°C
MDL nummer: MFCD00001602
CAS-nummer: 490-91-5
EINECS: 207-721-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=086f4e47d51d1d1993f7ddd4a28dcce107d93222&vid=086f4e47d51d1d1993f7ddd4a28dcce107d93222&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR