Triethylene glycol diacetate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Triethylene Glycol Diacetate

T0923-25G T0923-500G
TCIAT0923-25GEA 230 NOK
TCIAT0923-25G TCIAT0923-500G
Triethylene glycol diacetate ≥98.0% (by GC)
Triethylene glycol diacetate
Formel: C₁₀H₁₈O₆
MW: 234,25 g/mol
Kokepunkt: 300°C
Flash punkt: 174°C
MDL nummer: MFCD00026213
CAS-nummer: 111-21-7
EINECS: 203-846-0

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR