2,2,4-Trimethyl-1,3-pentanediol diisobutyrate ≥98.5% (by GC)

Leverandør: TCI

Synonymer: 2,2,4-Trimethylpentane-1,3-diyl bis(2-methylpropanoate)

T0997-25ML T0997-500ML
TCIAT0997-25MLEA 220 NOK
TCIAT0997-25ML TCIAT0997-500ML
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate ≥98.5% (by GC)
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate
Formel: C₁₆H₃₀O₄
MW: 286,41 g/mol
Kokepunkt: 280°C
Smeltepunkt: –70°C
Flash punkt: 121°C
MDL nummer: MFCD00059267
CAS-nummer: 6846-50-0
EINECS: 229-934-9
Merck Indeks: 14,09251

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR