3,3',5,5'-Tetra-tert-butyl-4,4'-stilbenequinone ≥95.0% (by HPLC)

Leverandør: TCI

T1067-1G T1067-5G
TCIAT1067-1GEA 2375 NOK
TCIAT1067-1G TCIAT1067-5G
3,3',5,5'-Tetra-tert-butyl-4,4'-stilbenequinone ≥95.0% (by HPLC)
3,3',5,5'-Tetra-tert-butyl-4,4'-stilbenequinone
Formel: C₃₀H₄₂O₂
MW: 434,66 g/mol
MDL nummer: MFCD00060073
CAS-nummer: 809-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44b4278a59877a090bbaaa43fc74e1674435dea7&vid=44b4278a59877a090bbaaa43fc74e1674435dea7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR