2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 2,2-Thiodiethylene Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]

T1070-25G T1070-500G T1070-100G
TCIAT1070-25GEA 340 NOK
TCIAT1070-25G TCIAT1070-500G TCIAT1070-100G
2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98.0% (by HPLC, titration analysis)
2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
Formel: C₃₈H₅₈O₆S
MW: 642,94 g/mol
MDL nummer: MFCD00059346
CAS-nummer: 41484-35-9
EINECS: 255-392-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e13991dcc4e2f62304c052cdc08c03341f487cdb&vid=e13991dcc4e2f62304c052cdc08c03341f487cdb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Saponification Value) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR