2-(3-Thienyl)pyridine ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-(2-Pyridyl)thiophene, 2-(Thiophen-3-yl)pyridine

T1151-5G T1151-1G
TCIAT1151-5GEA 8150 NOK
TCIAT1151-5G TCIAT1151-1G
2-(3-Thienyl)pyridine ≥95.0% (by GC)
2-(3-Thienyl)pyridine
Formel: C₉H₇NS
MW: 161,22 g/mol
MDL nummer: MFCD00006214
CAS-nummer: 21298-55-5
EINECS: 244-322-1

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR