Tris(4-bromophenyl)amine ≥98.0% (by GC)

Leverandør: TCI
Warning

T1361-25G T1361-5G
TCIAT1361-25GEA 1625 NOK
TCIAT1361-25G TCIAT1361-5G
Tris(4-bromophenyl)amine ≥98.0% (by GC)
Tris(4-bromophenyl)amine
Formel: C₁₈H₁₂Br₃N
MW: 482,01 g/mol
MDL nummer: MFCD00009665
CAS-nummer: 4316-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0752a0602cb3463cf697bc960ed0fae62498c8e&vid=a0752a0602cb3463cf697bc960ed0fae62498c8e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 144.0 to 147.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR