(3R)-(+)-3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI

T1369-1G T1369-25G
TCIAT1369-1GEA 650 NOK
TCIAT1369-1G TCIAT1369-25G
(3R)-(+)-3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)
(3R)-(+)-3-(Trifluoroacetamido)pyrrolidine hydrochloride
Formel: C₆H₉F₃N₂O
MW: 218,60 g/mol
MDL nummer: MFCD02094762
CAS-nummer: 141043-16-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +25.0 to +28.0 deg (C=1,H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR