Trilinolein ≥95.0% (by GC)

Leverandør: TCI

Synonymer: 1,2,3-Tri(cis,cis-9,12-octadecadienoyl) Glycerol, Glycerol Tri-9,12-octadecadienoate

T1388-25G T1388-5G
TCIAT1388-25GEA 7925 NOK
TCIAT1388-25G TCIAT1388-5G
Trilinolein ≥95.0% (by GC)
Trilinolein
Formel: C₅₇H₉₈O₆
MW: 879,41 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00042909
CAS-nummer: 537-40-6
EINECS: 208-666-6

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 0.9190 to 0.9290

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR