N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: TPEN, N1,N1,N2,N2-Tetrakis(pyridin-2-ylmethyl)ethane-1,2-diamine, Tetrakis-(2-pyridylmethyl)ethylenediamine

T1487-1G T1487-5G
TCIAT1487-1GEA 1500 NOK
TCIAT1487-1G TCIAT1487-5G
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine ≥97.0% (by titrimetric analysis)
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine
Formel: C₂₆H₂₈N₆
MW: 424,55 g/mol
MDL nummer: MFCD00036918
CAS-nummer: 16858-02-9

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR