Tris(dibenzoylmethanato)iron ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
Tris(1,3-diphenyl-1,3-propanedionato) Iron
T1686-25G
T1686-5G
TCIAT1686-25GEA
3325
NOK
TCIAT1686-25G
TCIAT1686-5G
Tris(dibenzoylmethanato)iron ≥97.0% (by titrimetric analysis)
Tris(dibenzoylmethanato)iron
Formel:
C₄₅H₃₃FeO₆ MW: 725,60 g/mol |
MDL nummer:
MFCD00192557 CAS-nummer: 14405-49-3 |
Spesifikasjon Testresultater
Appearance | Red to Dark blue to Black powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...