N,N',N''-Triphenyl-1,3,5-benzenetriamine ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
1,3,5-Tris(phenylamino)benzene
T1831-25G
T1831-5G
TCIAT1831-25GEA
4275
NOK
TCIAT1831-25G
TCIAT1831-5G
N,N',N''-Triphenyl-1,3,5-benzenetriamine ≥98.0% (by HPLC, titration analysis)
N,N',N''-Triphenyl-1,3,5-benzenetriamine
Formel:
C₂₄H₂₁N₃ MW: 351,45 g/mol |
MDL nummer:
MFCD01463903 CAS-nummer: 102664-66-4 |
Spesifikasjon Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...