[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: Cobalt tetramethoxyphenylporphyrin, [5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphinato]cobalt(II), Cobalt(II) meso-tetra(4-methoxyphenyl)porphine, 5,10,15,20-Tetrakis (4-methoxyphenyl)-21H,23H-porphine cobalt (II), 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)

T1861-1G T1861-5G
TCIAT1861-1GEA 755 NOK
TCIAT1861-1G TCIAT1861-5G
[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) ≥96.0% (by titrimetric analysis)
[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II)
Formel: C₄₈H₃₆CoN₄O₄
MW: 791.77 g/mol
MDL nummer: MFCD00010724
CAS-nummer: 28903-71-1

Bestill nå

Spesifikasjon Testresultater

Appearance Dark red to Dark purple to Dark blue powder to crystal
Purity (Chelometric Titration) min. 96.0 %
Lambda max. 415.0 to 425.0 nm (THF)
Absorbance (E1%1cm) min. 2000 (THF, 415.0 to 425.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR