3,4,5-Tris(benzyloxy)benzoic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3,4,5-Tribenzyloxybenzoic Acid

T1869-5G T1869-25G
TCIAT1869-5GEA 1925 NOK
TCIAT1869-5G TCIAT1869-25G
3,4,5-Tris(benzyloxy)benzoic acid ≥98.0% (by HPLC, titration analysis)
3,4,5-Tris(benzyloxy)benzoic acid
Formel: C₂₈H₂₄O₅
MW: 440,50 g/mol
MDL nummer: MFCD00825832
CAS-nummer: 1486-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4139e99d93d1d9f91ef50775d58a67c969f7b64&vid=b4139e99d93d1d9f91ef50775d58a67c969f7b64&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 193.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR